CAS RN 300657-03-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ FABP4 Inhibitor
The FABP4 Inhibitor, also referenced under CAS 300657-03-8, controls the biological activity of FABP4.