accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3007-31-6

IUPAC Name: didodecylamine
Synonyms: didodecylamine
Molecular Weight (g/mol): 353.68
Molecular Formula: C24H51N
InChi Key: MJCJUDJQDGGKOX-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCNCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)