CAS RN 3007-74-7
IUPAC Name:
N-dodecylaniline
Synonyms:
N-dodecylaniline
Molecular Weight (g/mol):
261.45
Molecular Formula:
C18H31N
InChi Key:
LQKYCMRSWKQVBQ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCNC1=CC=CC=C1
1
–
1
of
1
results
1
N-Dodecylaniline 93.0+%, TCI America™
CAS: 3007-74-7 Molecular Formula: C18H31N Molecular Weight (g/mol): 261.45 MDL Number: MFCD00026570 InChI Key: LQKYCMRSWKQVBQ-UHFFFAOYSA-N PubChem CID: 76362 IUPAC Name: N-dodecylaniline SMILES: CCCCCCCCCCCCNC1=CC=CC=C1