accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 300766-53-4

IUPAC Name: 4-methylbenzen-1-ide; iodozincylium
Synonyms: 4-methylbenzen-1-ide; iodozincylium
Molecular Weight (g/mol): 283.42
Molecular Formula: C7H7IZn
InChi Key: AHAJMZHMDYRVRB-UHFFFAOYSA-M
SMILES: [Zn+]I.CC1=CC=[C-]C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results