accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCZnI

CAS RN 300825-30-3

IUPAC Name: anisole; iodozincylium
Synonyms: anisole; iodozincylium
Molecular Weight (g/mol): 299.42
Molecular Formula: C7H7IOZn
InChi Key: GZPQUUMHBPXZTL-UHFFFAOYSA-M
SMILES: [Zn+]I.COC1=C[C-]=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results