accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 301-13-3

IUPAC Name: tris(2-ethylhexyl) phosphite
Synonyms: tri-(2-ethylhexyl)phosphite
Molecular Weight (g/mol): 418.64
Molecular Formula: C24H51O3P
InChi Key: ILLOBGFGKYTZRO-UHFFFAOYNA-N
SMILES: CCCCC(CC)COP(OCC(CC)CCCC)OCC(CC)CCCC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)