accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3010-82-0

IUPAC Name: benzene-1,4-dicarboxamide
Synonyms: terephthalamide
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: MHSKRLJMQQNJNC-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(C=C1)C(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results