accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 301177-50-4

IUPAC Name: N'-hydroxy-3-(2-methyl-4-nitro-1H-imidazol-1-yl)propanimidamide
Synonyms: N'-hydroxy-3-(2-methyl-4-nitroimidazol-1-yl)propanimidamide
Molecular Weight (g/mol): 213.2
Molecular Formula: C7H11N5O3
InChi Key: DSFFWFHDJMMTGE-UHFFFAOYSA-N
SMILES: CC1=NC(=CN1CCC(N)=NO)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results