accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3012-80-4

IUPAC Name: 1-methyl-1H-1,3-benzodiazole-2-carbaldehyde
Synonyms: 1-methyl-1,3-benzodiazole-2-carbaldehyde
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: SIRPHJCQZYVEES-UHFFFAOYSA-N
SMILES: CN1C(C=O)=NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results