CAS RN 301524-28-7
IUPAC Name:
calcium didihydrogen phosphate
Synonyms:
monocalcium phosphate|calcium bis(dihydrogenphosphate)
Molecular Weight (g/mol):
234.05
Molecular Formula:
CaH4O8P2
InChi Key:
YYRMJZQKEFZXMX-UHFFFAOYSA-L
SMILES:
[Ca++].OP(O)([O-])=O.OP(O)([O-])=O
1
–
1
of
1
results
1
Calcium dihydrogen phosphate hydrate, 97%
CAS: 301524-28-7 Molecular Formula: CaH4O8P2 Molecular Weight (g/mol): 234.05 MDL Number: MFCD00010898 InChI Key: YYRMJZQKEFZXMX-UHFFFAOYSA-L Synonym: Calcium phosphate, monobasic; Calcium tetrahydrogen diphosphate IUPAC Name: calcium didihydrogen phosphate SMILES: [Ca++].OP(O)([O-])=O.OP(O)([O-])=O