accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 3016-97-5

IUPAC Name: (4-benzoylphenyl)(phenyl)methanone
Synonyms: (4-benzoylphenyl)(phenyl)methanone
Molecular Weight (g/mol): 286.33
Molecular Formula: C20H14O2
InChi Key: NPENBPVOAXERED-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results