accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30165-96-9

IUPAC Name: 4-(4-chloro-1,2,5-thiadiazol-3-yl)morpholine
Synonyms: 4-(4-chloro-1,2,5-thiadiazol-3-yl)morpholine
Molecular Weight (g/mol): 205.66
Molecular Formula: C6H8ClN3OS
InChi Key: LAUCCQWGVCJGFT-UHFFFAOYSA-N
SMILES: ClC1=NSN=C1N1CCOCC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)