accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3019-25-8

IUPAC Name: 1-cyclobutylethan-1-one
Synonyms: ethanone, 1-cyclobutyl-
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: JPJOOTWNILDNAW-UHFFFAOYSA-N
SMILES: CC(=O)C1CCC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)