accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3019-58-7

IUPAC Name: 2,3-dihydroxy-3-(phenylcarbamoyl)propanoic acid
Synonyms: 2,3-dihydroxy-3-(phenylcarbamoyl)propanoic acid
Molecular Weight (g/mol): 225.2
Molecular Formula: C10H11NO5
InChi Key: ZWXNRJCDXZFNLJ-UHFFFAOYNA-N
SMILES: OC(C(O)C(=O)NC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results