accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHNH3OHSOOO

CAS RN 302-15-8

IUPAC Name: 2-methylhydrazinium hydrogen sulfate
Synonyms: 2-methylhydrazinium hydrogen sulfate
Molecular Weight (g/mol): 144.15
Molecular Formula: CH8N2O4S
InChi Key: KJDJPXUIZYHXEZ-UHFFFAOYSA-N
SMILES: CN[NH3+].OS([O-])(=O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)