accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHOHO

CAS RN 3025-96-5

IUPAC Name: 4-acetamidobutanoic acid
Synonyms: 4-acetamidobutyrate 4-(acetylamino)butyrate
Molecular Weight (g/mol): 145.16
Molecular Formula: C6H11NO3
InChi Key: UZTFMUBKZQVKLK-UHFFFAOYSA-N
SMILES: CC(=O)NCCCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)