accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 302554-80-9

IUPAC Name: 2-bromo-9,9-dioctyl-9H-fluorene
Synonyms: 2-bromo-9,9-dioctylfluorene
Molecular Weight (g/mol): 469.55
Molecular Formula: C29H41Br
InChi Key: ITVGRPGDCPNGHZ-UHFFFAOYSA-N
SMILES: CCCCCCCCC1(CCCCCCCC)C2=CC=CC=C2C2=C1C=C(Br)C=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results