accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3COSOOO

CAS RN 3026-63-9

IUPAC Name: sodium tridecyl sulfate
Synonyms: sodium tridecyl sulfate
Molecular Weight (g/mol): 302.4
Molecular Formula: C13H27NaO4S
InChi Key: HQCFDOOSGDZRII-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCCCCOS([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results