accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3027-13-2

IUPAC Name: 1-(3-methoxyphenyl)propan-2-one
Synonyms: 3-methoxyphenyl methyl ketone
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: RMMRRRLPDBJBQL-UHFFFAOYSA-N
SMILES: COC1=CC=CC(CC(C)=O)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results