accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSiH3COCH3

CAS RN 3027-21-2

IUPAC Name: dimethoxy(methyl)phenylsilane
Synonyms: methylphenyldimethoxysilane
Molecular Weight (g/mol): 182.29
Molecular Formula: C9H14O2Si
InChi Key: CVQVSVBUMVSJES-UHFFFAOYSA-N
SMILES: CO[Si](C)(OC)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)