accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClClH2N

CAS RN 30273-00-8

IUPAC Name: 2,4-dichloro-6-methylaniline
Synonyms: 2,4-dichloro-6-methylaniline
Molecular Weight (g/mol): 176.04
Molecular Formula: C7H7Cl2N
InChi Key: UAISVUGQLKXPFF-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC(Cl)=C1N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)