accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30289-28-2

IUPAC Name: N-methoxy-N-methylcarbamoyl chloride
Synonyms: N-methoxy-N-methylcarbamoyl chloride
Molecular Weight (g/mol): 123.54
Molecular Formula: C3H6ClNO2
InChi Key: SHQJHXLWZDMQEJ-UHFFFAOYSA-N
SMILES: CON(C)C(Cl)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)