accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3029-32-1

IUPAC Name: 6,13-dihydropentacene-6,13-dione
Synonyms: pentacene-6,13-dione
Molecular Weight (g/mol): 308.34
Molecular Formula: C22H12O2
InChi Key: UFCVADNIXDUEFZ-UHFFFAOYSA-N
SMILES: O=C1C2=C(C=C3C=CC=CC3=C2)C(=O)C2=C1C=C1C=CC=CC1=C2
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)