accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFBrFFF

CAS RN 302911-98-4

IUPAC Name: 2-(bromomethyl)-1,4-bis(trifluoromethyl)benzene
Synonyms: 2-(bromomethyl)-1,4-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 307.03
Molecular Formula: C9H5BrF6
InChi Key: CHSQGVCRNVEWIA-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C(CBr)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results