accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClNHONSH2N

CAS RN 302964-24-5

IUPAC Name: 2-amino-N-(2-chloro-6-methylphenyl)-1,3-thiazole-5-carboxamide
Synonyms: 2-amino-N-(2-chloro-6-methylphenyl)-1,3-thiazole-5-carboxamide
Molecular Weight (g/mol): 267.73
Molecular Formula: C11H10ClN3OS
InChi Key: VVOXTERFTAJMAA-UHFFFAOYSA-N
SMILES: CC1=C(NC(=O)C2=CN=C(N)S2)C(Cl)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results