accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30299-08-2

CH3H3COOH3CCH3OHOOHO

CAS RN 30299-08-2

IUPAC Name: 2-(4-{1-[4-(1-carboxy-1-methylpropoxy)phenyl]cyclohexyl}phenoxy)-2-methylbutanoic acid
Synonyms: clinofibrate (R*,S*)-clinofibrate (r*,s*)-clinofibrate clinofibrate, (r*,s*)- (R*,S*)-isomer clinofibrate clinofibrate, (r*,s*)-isomer clinofibrato (r*,s*)-isomer clinofibrate 克利贝特 clinofibratum
Molecular Weight (g/mol): 468.59
Molecular Formula: C28H36O6
InChi Key: BMOVQUBVGICXQN-UHFFFAOYNA-N
SMILES: CCC(C)(OC1=CC=C(C=C1)C1(CCCCC1)C1=CC=C(OC(C)(CC)C(O)=O)C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results