accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 303-47-9

IUPAC Name: (2S)-2-{[(3R)-5-chloro-8-hydroxy-3-methyl-1-oxo-3,4-dihydro-1H-2-benzopyran-7-yl]formamido}-3-phenylpropanoic acid
Synonyms: ochratoxin A
Molecular Weight (g/mol): 403.82
Molecular Formula: C20H18ClNO6
InChi Key: RWQKHEORZBHNRI-BMIGLBTASA-N
SMILES: C[C@@H]1CC2=C(Cl)C=C(C(=O)N[C@@H](CC3=CC=CC=C3)C(O)=O)C(O)=C2C(=O)O1
Percent Purity 
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)