accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NCH3NCH3CH3

CAS RN 3030-47-5

IUPAC Name: (2-{[2-(dimethylamino)ethyl](methyl)amino}ethyl)dimethylamine
Synonyms: pentamethyldiethylenetriamine pmdien pentamethyldiethylenetriaminek pmdta PMDT
Molecular Weight (g/mol): 173.3
Molecular Formula: C9H23N3
InChi Key: UKODFQOELJFMII-UHFFFAOYSA-N
SMILES: CN(C)CCN(C)CCN(C)C
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (6)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)