accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOCH3OOCH3OOOCH3

CAS RN 303127-79-9

IUPAC Name: [3,5-bis(acetyloxy)-4-(benzyloxy)-6-(4-methoxyphenoxy)oxan-2-yl]methyl acetate
Synonyms: [3,5-bis(acetyloxy)-4-(benzyloxy)-6-(4-methoxyphenoxy)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 502.52
Molecular Formula: C26H30O10
InChi Key: MDZXNXKMNWQSDZ-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COC(C)=O)C(OC(C)=O)C(OCC3=CC=CC=C3)C2OC(C)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)