accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30315-46-9

IUPAC Name: tert-butyl[(2S)-2,3-dihydroxypropyl]azanium
Synonyms: tert-butyl[(2S)-2,3-dihydroxypropyl]azanium
Molecular Weight (g/mol): 148.23
Molecular Formula: C7H18NO2
InChi Key: JWBMVCAZXJMSOX-LURJTMIESA-O
SMILES: CC(C)(C)[NH2+]C[C@H](O)CO
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results