accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CFOFFFFFF

CAS RN 303186-19-8

IUPAC Name: 4-[difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluoro-1,1'-biphenyl
Synonyms: 4-[difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluoro-1,1'-biphenyl
Molecular Weight (g/mol): 414.32
Molecular Formula: C21H13F7O
InChi Key: ZTUBOAYBUBUTPH-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)C1=CC(F)=C(C(F)=C1)C(F)(F)OC1=CC(F)=C(F)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results