CAS RN 3032-32-4
IUPAC Name:
2-amino-3,6-dichlorobenzoic acid
Synonyms:
2-amino-3,6-dichlorobenzoic acid
Molecular Weight (g/mol):
206.02
Molecular Formula:
C7H5Cl2NO2
InChi Key:
MMIREQFMYZQWLU-UHFFFAOYSA-N
SMILES:
NC1=C(Cl)C=CC(Cl)=C1C(O)=O
1
–
1
of
1
results
1
Promotions Available
2-Amino-3,6-dichlorobenzoic acid, 95%, Thermo Scientific Chemicals
2-Amino-3,6-dichlorobenzoic acid, 95%, Thermo Scientific Chemicals
CAS: 3032-32-4 Molecular Formula: C7H5Cl2NO2 Molecular Weight (g/mol): 206.02 MDL Number: MFCD00052390 InChI Key: MMIREQFMYZQWLU-UHFFFAOYSA-N PubChem CID: 604542 IUPAC Name: 2-amino-3,6-dichlorobenzoic acid SMILES: NC1=C(Cl)C=CC(Cl)=C1C(O)=O