accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30322-02-2

IUPAC Name: 7-methyl-3,4-dihydro-2H-1,4-benzothiazin-3-one
Synonyms: 7-methyl-2,4-dihydro-1,4-benzothiazin-3-one
Molecular Weight (g/mol): 179.24
Molecular Formula: C9H9NOS
InChi Key: ODJQMWDFNLNCTM-UHFFFAOYSA-N
SMILES: CC1=CC2=C(NC(=O)CS2)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results