accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3033-80-5

H3COCH3N

CAS RN 3033-80-5

IUPAC Name: 8-methoxy-2-methylquinoline
Synonyms: 8-methoxy-2-methylquinoline
Molecular Weight (g/mol): 173.22
Molecular Formula: C11H11NO
InChi Key: OQXVXPBDSJQYRR-UHFFFAOYSA-N
SMILES: COC1=C2N=C(C)C=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results