accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30339-30-1

IUPAC Name: 2-(4-methylphenyl)-1-phenylethan-1-amine
Synonyms: 2-(4-methylphenyl)-1-phenylethanamine
Molecular Weight (g/mol): 211.31
Molecular Formula: C15H17N
InChi Key: ZICDZTXDTPZBKH-UHFFFAOYNA-N
SMILES: CC1=CC=C(CC(N)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results