accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3034-34-2

IUPAC Name: 4-cyanobenzamide
Synonyms: P-cyanobenzamide
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: FUKWTMJZHKZKFA-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results