accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONHN

CAS RN 3034-38-6

IUPAC Name: 5-nitro-1H-imidazole
Synonyms: 5-nitroimidazole
Molecular Weight (g/mol): 113.08
Molecular Formula: C3H3N3O2
InChi Key: VYDWQPKRHOGLPA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=CN1
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (7)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)