accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSN

CAS RN 3034-52-4

IUPAC Name: 2-chloro-1,3-thiazole
Synonyms: 2-chloro-1,3-thiazole C3H2ClNS
Molecular Weight (g/mol): 119.57
Molecular Formula: C3H2ClNS
InChi Key: KLEYVGWAORGTIT-UHFFFAOYSA-N
SMILES: ClC1=NC=CS1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)