accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3034-53-5

IUPAC Name: 2-bromo-1,3-thiazole
Synonyms: thiazole, 2-bromo-
Molecular Weight (g/mol): 164.02
Molecular Formula: C3H2BrNS
InChi Key: RXNZFHIEDZEUQM-UHFFFAOYSA-N
SMILES: BrC1=NC=CS1
Boiling Point 
  • (5)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (5)
  • (1)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)