accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HS

CAS RN 3037-04-5

IUPAC Name: 2-aminoethane-1-thiol
Synonyms: cysteamine riacon mercaptamine mercamine cysteamin cysteinamine cysteamide becaptan bitartrate, cysteamine cysteamine (product)
Molecular Weight (g/mol): 77.15
Molecular Formula: C2H7NS
InChi Key: UFULAYFCSOUIOV-UHFFFAOYSA-N
SMILES: NCCS
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results