accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNNClOH

CAS RN 303727-31-3

IUPAC Name: 2-chloro-5-[2-phenyl-5-(pyridin-4-yl)-1H-imidazol-4-yl]phenol
Synonyms: 2-chloro-5-[2-phenyl-5-(pyridin-4-yl)-1H-imidazol-4-yl]phenol
Molecular Weight (g/mol): 347.8
Molecular Formula: C20H14ClN3O
InChi Key: WXJLXRNWMLWVFB-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=CC(=C1)C1=C(NC(=N1)C1=CC=CC=C1)C1=CC=NC=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results