CAS RN 303997-35-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ TNF-α Antagonist III, R-7050
The TNF-α Antagonist III, R-7050, also referenced under CAS 303997-35-5, controls the biological activity of TNF-α.