accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 304-17-6

IUPAC Name: 2-(propan-2-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: isopropylphthalimide
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: VPLDXHDOGVIETL-UHFFFAOYSA-N
SMILES: CC(C)N1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results