accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 304-91-6

IUPAC Name: 2-iodosylbenzoic acid
Synonyms: o-iodosobenzoic acid
Molecular Weight (g/mol): 264.02
Molecular Formula: C7H5IO3
InChi Key: IFPHDUVGLXEIOQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1[I]=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)