accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3041-16-5

IUPAC Name: 1,4-dioxan-2-one
Synonyms: 1,4-dioxan-2-one
Molecular Weight (g/mol): 102.09
Molecular Formula: C4H6O3
InChi Key: VPVXHAANQNHFSF-UHFFFAOYSA-N
SMILES: O=C1COCCO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results