accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3042-00-0

IUPAC Name: 2-(1H-1,3-benzodiazol-2-ylsulfanyl)acetate
Synonyms: 2-(1H-1,3-benzodiazol-2-ylsulfanyl)acetate
Molecular Weight (g/mol): 207.23
Molecular Formula: C9H7N2O2S
InChi Key: UYNVBLJQBCTRKV-UHFFFAOYSA-M
SMILES: [O-]C(=O)CSC1=NC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results