accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3OCH3OH3C

CAS RN 3044-56-2

IUPAC Name: ethyl 3-oxo-3-(3,4,5-trimethoxyphenyl)propanoate
Synonyms: ethyl 3-oxo-3-(3,4,5-trimethoxyphenyl)propanoate
Molecular Weight (g/mol): 282.29
Molecular Formula: C14H18O6
InChi Key: VEYNISLTHXQSGU-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC(OC)=C(OC)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results