accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30459-17-7

IUPAC Name: 1-[4-(trifluoromethyl)phenyl]piperazine
Synonyms: 1-[4-(trifluoromethyl)phenyl]piperazine
Molecular Weight (g/mol): 230.23
Molecular Formula: C11H13F3N2
InChi Key: IBQMAPSJLHRQPE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)N1CCNCC1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)