accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOH2ONa

CAS RN 304672-01-3

IUPAC Name: sodium hydrate propane-1-sulfonate
Synonyms: sodium hydrate propane-1-sulfonate
Molecular Weight (g/mol): 164.15
Molecular Formula: C3H9NaO4S
InChi Key: QBQVXXQXZXDEHE-UHFFFAOYSA-M
SMILES: O.[Na+].CCCS([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results