accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOH3COOH3COOH3CSc

CAS RN 304675-64-7

IUPAC Name: scandium(3+) triacetate
Synonyms: scandium(3+) triacetate
Molecular Weight (g/mol): 222.09
Molecular Formula: C6H9O6Sc
InChi Key: DBTMQFKUVICLQN-UHFFFAOYSA-K
SMILES: [Sc+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results